Gene Transcripts

Gene Symbol
Krt16
Entrez Gene ID
303530
Gene Name
keratin 16, type I
Gene Aliases
Ka16
Chromosome Location
Chr. 10: 88149210 - 88152142 on Build Rnor_6.0
UniGene ID
Rn.145931
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008752.1 NP_001008752.1 2, 3 583
XM_006247309.2 XP_006247371.1 2, 3 661

View Full Product Details