Gene Transcripts

Gene Symbol
Nags
Entrez Gene ID
303563
Gene Name
N-acetylglutamate synthase
Gene Aliases
RGD1565783
Chromosome Location
Chr. 10: 90084609 - 90089693 on Build Rnor_6.0
UniGene ID
Rn.23912
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107053.1 NP_001100523.1 3 931
XM_006247328.2 XP_006247390.1 4 1751
XM_008768055.1 XP_008766277.1 4 1751

View Full Product Details