Gene Transcripts

Gene Symbol
Lyl1
Entrez Gene ID
304663
Gene Name
lymphoblastic leukemia associated hematopoiesis regulator 1
Gene Aliases
Chromosome Location
Chr. 19: 25815207 - 25818074 on Build Rnor_6.0
UniGene ID
Rn.17352
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007677.1 NP_001007678.1 2, 3 550
GenBank mRNA BC081864.1 - 2, 3 550

View Full Product Details