Gene Transcripts

Gene Symbol
Fancl
Entrez Gene ID
305600
Gene Name
Fanconi anemia, complementation group L
Gene Aliases
Chromosome Location
Chr. 14: 110675322 - 110740880 on Build Rnor_6.0
UniGene ID
Rn.82534
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191684.1 NP_001178613.1 12 922
XM_006251597.2 XP_006251659.1 11 1621

View Full Product Details