Gene Transcripts
- Gene Symbol
- Mettl17
- Entrez Gene ID
- 305845
- Gene Name
- methyltransferase like 17
- Gene Aliases
- Mett11d1, RGD1559895
- Chromosome Location
- Chr. 15: 28287033 - 28293869 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_223974.8 |
XP_223974.4
|
8, 9 |
1017 |
|
View Full Product Details