Gene Transcripts

Gene Symbol
Cenpj
Entrez Gene ID
305909
Gene Name
centromere protein J
Gene Aliases
Chromosome Location
Chr. 15: 36745672 - 36807551 on Build Rnor_6.0
UniGene ID
Rn.19524
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107265.1 NP_001100735.1 6 1832
XM_008770747.1 XP_008768969.1 7 5270

View Full Product Details