Gene Transcripts

Gene Symbol
Lgi3
Entrez Gene ID
306013
Gene Name
leucine-rich repeat LGI family, member 3
Gene Aliases
Chromosome Location
Chr. 15: 52220578 - 52227713 on Build Rnor_6.0
UniGene ID
Rn.50862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107277.1 NP_001100747.1 8 1373
XM_006252273.2 XP_006252335.1 7 1292

View Full Product Details