Gene Transcripts

Gene Symbol
Fam160b2
Entrez Gene ID
306015
Gene Name
family with sequence similarity 160, member B2
Gene Aliases
Rai16
Chromosome Location
Chr. 15: 52271495 - 52288893 on Build Rnor_6.0
UniGene ID
Rn.124637
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170474.1 NP_001163945.1 7, 8 991
XM_006252274.2 XP_006252336.1 7, 8 1488
GenBank mRNA BC168691.1 - 7, 8 991

View Full Product Details