Gene Transcripts

Gene Symbol
Myom2
Entrez Gene ID
306616
Gene Name
myomesin 2
Gene Aliases
Chromosome Location
Chr. 16: 79587136 - 79672847 on Build Rnor_6.0
UniGene ID
Rn.33916
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169141.1 NP_001162612.1 23 3104
XM_006253389.2 XP_006253451.1 23 3205
XM_008771360.1 XP_008769582.1 23 3022

View Full Product Details