Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004720.2 | NP_001004720.1 | 4 | 2004 | |
NM_001004722.3 | NP_001004722.1 | 4 | 1392 | ||
NM_003581.4 | NP_003572.2 | 5 | 2114 | ||
XM_006712797.3 | XP_006712860.1 | 5 | 2770 | ||
XM_011511991.2 | XP_011510293.1 | 5 | 2208 | ||
XM_011511992.2 | XP_011510294.2 | 3 | 1336 | ||
XM_017005103.1 | XP_016860592.1 | 6 | 2372 | ||
XM_017005104.1 | XP_016860593.1 | 5 | 2384 | ||
XM_017005105.1 | XP_016860594.1 | 5 | 2121 | ||
GenBank mRNA | BC000103.2 | - | 4 | 1892 | |
BC007195.1 | - | 4 | 1899 | 3608149 |