Gene Transcripts

Gene Symbol
Mocs3
Entrez Gene ID
311655
Gene Name
molybdenum cofactor synthesis 3
Gene Aliases
Chromosome Location
Chr. 3: 164986421 - 164988381 on Build Rnor_6.0
UniGene ID
Rn.35140
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107804.1 NP_001101274.1 1 1204

View Full Product Details