Gene Transcripts

Gene Symbol
Zc3h12a
Entrez Gene ID
313587
Gene Name
zinc finger CCCH type containing 12A
Gene Aliases
RGD1306776
Chromosome Location
Chr. 5: 143111339 - 143120131 on Build Rnor_6.0
UniGene ID
Rn.234344
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077671.1 NP_001071139.1 2 364
XM_006238865.2 XP_006238927.1 1 994
GenBank mRNA BC127516.1 - 2 364

View Full Product Details