Gene Transcripts

Gene Symbol
Dpy19l1
Entrez Gene ID
315496
Gene Name
dpy-19-like 1 (C. elegans)
Gene Aliases
RGD1305822
Chromosome Location
Chr. 8: 25582938 - 25669478 on Build Rnor_6.0
UniGene ID
Rn.22725
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191791.1 NP_001178720.1 10 1033
XM_006242719.2 XP_006242781.1 10 1117

View Full Product Details