Gene Transcripts

Gene Symbol
Pcdh11x
Entrez Gene ID
317204
Gene Name
protocadherin 11 X-linked
Gene Aliases
RGD1562864
Chromosome Location
Chr. X: 92596314 - 93341412 on Build Rnor_6.0
UniGene ID
Rn.234112
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271228.1 NP_001258157.1 3 2318
XM_008773375.1 XP_008771597.1 3 2365

View Full Product Details