Gene Transcripts

Gene Symbol
Spin2a
Entrez Gene ID
317395
Gene Name
spindlin family, member 2A
Gene Aliases
RGD1560898, Spin2
Chromosome Location
Chr. X: 17962905 - 17964888 on Build Rnor_6.0
UniGene ID
Rn.203523
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127657.2 NP_001121129.2 2 799
GenBank mRNA BC158655.1 - 2 799

Gene Symbol
LOC100909485
Entrez Gene ID
100909485
Gene Name
spindlin-2-like
Gene Aliases
Chromosome Location
Chr. X: 18340413 - 18342430 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003752019.3 XP_003752067.1 2 833

View Full Product Details