Gene Transcripts

Gene Symbol
Lhfpl4
Entrez Gene ID
353230
Gene Name
lipoma HMGIC fusion partner-like 4
Gene Aliases
Chromosome Location
Chr. 4: 145117533 - 145147397 on Build Rnor_6.0
UniGene ID
Rn.145133
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181387.2 NP_852052.1 2 519
GenBank mRNA AY278321.1 - 2 519

View Full Product Details