Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020981.3 | NP_066191.1 | 2 | 1101 | |
XM_005246931.3 | XP_005246988.1 | 5 | 1535 | ||
XM_006712819.3 | XP_006712882.1 | 3 | 5809 | ||
XM_011512085.2 | XP_011510387.1 | 5 | 1551 | ||
GenBank mRNA | AF117222.1 | - | 1 | 842 | |
BC101545.1 | - | 1 | 801 | 8069051 | |
BC104813.1 | - | 1 | 801 | 8143816 | |
E07739.1 | - | 5 | 1425 |