Gene Transcripts

Gene Symbol
Spag8
Entrez Gene ID
362508
Gene Name
sperm associated antigen 8
Gene Aliases
Chromosome Location
Chr. 5: 59147412 - 59149625 on Build Rnor_6.0
UniGene ID
Rn.144402
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173555.1 NP_001167026.1 2 326
GenBank mRNA BC079031.1 - 2 326

View Full Product Details