Gene Transcripts
- Gene Symbol
- Zyg11b
- Entrez Gene ID
- 362559
- Gene Name
- zyg-11 family member B, cell cycle regulator
- Gene Aliases
- RGD1307814
- Chromosome Location
- Chr. 5: 127840588 - 127878796 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_342878.8 |
XP_342879.3
|
1 |
292 |
|
View Full Product Details