Gene Transcripts

Gene Symbol
Glrx5
Entrez Gene ID
362776
Gene Name
glutaredoxin 5
Gene Aliases
RGD1308383
Chromosome Location
Chr. 6: 128750503 - 128760880 on Build Rnor_6.0
UniGene ID
Rn.104008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108722.1 NP_001102192.1 1, 2 317

View Full Product Details