Gene Transcripts

Gene Symbol
Pcdha2
Entrez Gene ID
393086
Gene Name
protocadherin alpha 2
Gene Aliases
rCNRv02
Chromosome Location
Chr. 18: 29960072 - 30215896 on Build Rnor_6.0
UniGene ID
Rn.22308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199504.1 NP_955798.1 1 753
GenBank mRNA AB113385.1 - 1 753
AY573976.1 - 1 753

View Full Product Details