Gene Transcripts

Gene Symbol
Jph4
Entrez Gene ID
445271
Gene Name
junctophilin 4
Gene Aliases
Chromosome Location
Chr. 15: 33767116 - 33776447 on Build Rnor_6.0
UniGene ID
Rn.163297
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003711.1 NP_001003711.1 3 1256
XM_008770690.1 XP_008768912.1 4 2054
GenBank mRNA AY341260.1 - 3 1256

View Full Product Details