Gene Transcripts

Gene Symbol
Vom1r63
Entrez Gene ID
494311
Gene Name
vomeronasal 1 receptor 63
Gene Aliases
V1re15
Chromosome Location
Chr. 1: 68670127 - 68671059 on Build Rnor_6.0
UniGene ID
Rn.158179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008967.1 NP_001008967.1 1 524
GenBank mRNA AY510345.1 - 1 524

View Full Product Details