Gene Transcripts

Gene Symbol
Kctd2
Entrez Gene ID
498024
Gene Name
potassium channel tetramerization domain containing 2
Gene Aliases
RGD1566063
Chromosome Location
Chr. 10: 103972750 - 103984398 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768411.1 XP_008766633.1 6 904

View Full Product Details