Gene Transcripts

Gene Symbol
Vom2r58
Entrez Gene ID
501793
Gene Name
vomeronasal 2 receptor, 58
Gene Aliases
Chromosome Location
Chr. 12: 5057134 - 5201595 on Build Rnor_6.0
UniGene ID
Rn.217929
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099479.1 NP_001092949.1 2 451

View Full Product Details