Gene Transcripts

Gene Symbol
Sult6b1
Entrez Gene ID
503103
Gene Name
sulfotransferase family, cytosolic, 6B, member 1
Gene Aliases
Chromosome Location
Chr. 6: 1468083 - 1493670 on Build Rnor_6.0
UniGene ID
Rn.155063
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001192017.1 NP_001178946.1 2 263

View Full Product Details