Gene Transcripts

Gene Symbol
Crisp4
Entrez Gene ID
654517
Gene Name
cysteine-rich secretory protein 4
Gene Aliases
Chromosome Location
Chr. 9: 24356577 - 24383679 on Build Rnor_6.0
UniGene ID
Rn.161107
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039393.2 NP_001034482.2 8 815
XM_008766935.1 XP_008765157.1 8 815
GenBank mRNA DQ200907.1 - 8 815

View Full Product Details