Gene Transcripts

Gene Symbol
Chmp4bl1
Entrez Gene ID
679886
Gene Name
chromatin modifying protein 4B-like 1
Gene Aliases
Chromosome Location
Chr. 4: 67059939 - 67060988 on Build Rnor_6.0
UniGene ID
Rn.234608
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276456.1 NP_001263385.1 1 1002

View Full Product Details