Gene Transcripts

Gene Symbol
Mrpl20
Entrez Gene ID
680747
Gene Name
mitochondrial ribosomal protein L20
Gene Aliases
Chromosome Location
Chr. 5: 173248245 - 173252775 on Build Rnor_6.0
UniGene ID
Rn.2913
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109428.1 NP_001102898.1 2, 3 334
GenBank mRNA BC166541.1 - 2, 3 219
FQ224518.1 - 2, 3 238

View Full Product Details