Gene Transcripts

Gene Symbol
Zcwpw2
Entrez Gene ID
681381
Gene Name
zinc finger, CW type with PWWP domain 2
Gene Aliases
Chromosome Location
Chr. 8: 126268760 - 126402046 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766697.1 XP_008764919.1 4 1390

View Full Product Details