Gene Transcripts

Gene Symbol
Magel2
Entrez Gene ID
679875
Gene Name
melanoma antigen family L2
Gene Aliases
Chromosome Location
Chr. 1: 123015408 - 123019945 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001054803.5 XP_001054803.2 1 2869

View Full Product Details