Gene Transcripts

Gene Symbol
Msrb1
Entrez Gene ID
685059
Gene Name
methionine sulfoxide reductase B1
Gene Aliases
Sepx1, selX
Chromosome Location
Chr. 10: 14105750 - 14111463 on Build Rnor_6.0
UniGene ID
Rn.198860
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044285.2 NP_001037750.2 2 259
GenBank mRNA BC094309.1 - 2 249
CB812976.1 - 2 259
FQ220312.1 - 2 272

View Full Product Details