Gene Transcripts

Gene Symbol
Ankrd50
Entrez Gene ID
294988
Gene Name
ankyrin repeat domain 50
Gene Aliases
RGD1311665
Chromosome Location
Chr. 2: 125289071 - 125323528 on Build Rnor_6.0
UniGene ID
Rn.7482
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760920.1 XP_008759142.1 1, 2 160

View Full Product Details