Gene Transcripts

Gene Symbol
Cby3
Entrez Gene ID
690544
Gene Name
chibby homolog 3 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 10: 35855320 - 35860266 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767754.1 XP_008765976.1 2 534

View Full Product Details