Gene Transcripts

Gene Symbol
Pcp4l1
Entrez Gene ID
685448
Gene Name
Purkinje cell protein 4-like 1
Gene Aliases
Chromosome Location
Chr. 13: 89542377 - 89565813 on Build Rnor_6.0
UniGene ID
Rn.155340
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126093.2 NP_001119565.1 3 586
XM_008769760.1 XP_008767982.1 3 1427
GenBank mRNA BC086330.1 - 3 586
BC162039.1 - 3 428 7099690

View Full Product Details