Gene Transcripts

Gene Symbol
Gstt4
Entrez Gene ID
686922
Gene Name
glutathione S-transferase, theta 4
Gene Aliases
Chromosome Location
Chr. 20: 13778185 - 13784813 on Build Rnor_6.0
UniGene ID
Rn.190813
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109675.1 NP_001103145.1 3, 4 443
GenBank mRNA BC097423.1 - 3, 4 427

View Full Product Details