Gene Transcripts

Gene Symbol
Msrb3
Entrez Gene ID
680036
Gene Name
methionine sulfoxide reductase B3
Gene Aliases
Chromosome Location
Chr. 7: 62850506 - 62972480 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001055430.5 XP_001055430.4 7 1067
XM_006241414.2 XP_006241476.1 8 661
XM_008765427.1 XP_008763649.1 7 1087
XM_008765428.1 XP_008763650.1 8 711

View Full Product Details