Gene Transcripts

Gene Symbol
Cib4
Entrez Gene ID
688819
Gene Name
calcium and integrin binding family member 4
Gene Aliases
Chromosome Location
Chr. 6: 27241804 - 27303617 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001068424.4 XP_001068424.2 6 701

View Full Product Details