Gene Transcripts

Gene Symbol
Rhox13
Entrez Gene ID
691244
Gene Name
Reproductive homeobox 13
Gene Aliases
Chromosome Location
Chr. X: 124267995 - 124274290 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001077350.5 XP_001077350.3 1 346

View Full Product Details