Gene Transcripts

Gene Symbol
Spata13
Entrez Gene ID
305938
Gene Name
spermatogenesis associated 13
Gene Aliases
Chromosome Location
Chr. 15: 40937658 - 41066645 on Build Rnor_6.0
UniGene ID
Rn.100403
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191686.1 NP_001178615.1 1, 2 111
XM_006252113.2 XP_006252175.1 1, 2 184

View Full Product Details