Gene Transcripts

Gene Symbol
KCNK15
Entrez Gene ID
60598
Gene Name
potassium two pore domain channel subfamily K member 15
Gene Aliases
K2p15.1, KCNK11, KCNK14, KT3.3, TASK-5, TASK5, dJ781B1.1
Chromosome Location
Chr. 20: 44745605 - 44752313 on Build GRCh38
UniGene ID
Hs.528664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022358.3 NP_071753.2 2 1233
GenBank mRNA AF294350.1 - 2 1169
AF294351.1 - 2 1169
BC082981.1 - 1 21

View Full Product Details