Gene Transcripts
- Gene Symbol
- CTAG1A
- Entrez Gene ID
- 246100
- Gene Name
- cancer/testis antigen 1A
- Gene Aliases
- CT6.1, ESO1, LAGE-2, LAGE2A, NY-ESO-1
- Chromosome Location
- Chr. X: 154585154 - 154586816 on Build GRCh38
- UniGene ID
- Hs.534310
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_139250.1 |
NP_640343.1
|
2 |
435 |
|
- Gene Symbol
- CTAG1B
- Entrez Gene ID
- 1485
- Gene Name
- cancer/testis antigen 1B
- Gene Aliases
- CT6.1, CTAG, CTAG1, ESO1, LAGE-2, LAGE2B, NY-ESO-1
- Chromosome Location
- Chr. X: 154617609 - 154619271 on Build GRCh38
- UniGene ID
- Hs.534310
- Species
- Homo sapiens
View Full Product Details