Gene Transcripts

Gene Symbol
KRTAP12-2
Entrez Gene ID
353323
Gene Name
keratin associated protein 12-2
Gene Aliases
KAP12.2, KRTAP12.2
Chromosome Location
Chr. 21: 44666189 - 44666927 on Build GRCh38
UniGene ID
Hs.567902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181684.2 NP_859012.1 1 595
GenBank mRNA AB076362.1 - 1 574
AJ566389.1 - 1 604

View Full Product Details