Gene Transcripts
- Gene Symbol
- VKORC1L1
- Entrez Gene ID
- 154807
- Gene Name
- vitamin K epoxide reductase complex subunit 1 like 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 65835816 - 65959563 on Build GRCh38
- UniGene ID
- Hs.427232
- Species
- Homo sapiens
- Gene Symbol
- CCT6P3
- Entrez Gene ID
- 643180
- Gene Name
- chaperonin containing TCP1 subunit 6 pseudogene 3
- Gene Aliases
- CCT6-3P
- Chromosome Location
- Chr. 7: 65038354 - 65074713 on Build GRCh38
- UniGene ID
- Hs.510847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033416.1 |
|
1 |
109 |
|
GenBank mRNA |
BX649060.1 |
- |
1 |
110 |
|
View Full Product Details