Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 801 | |
XM_006716463.2 | XP_006716526.1 | 7 | 1394 | ||
GenBank mRNA | AK225246.1 | - | 1 | 800 | |
AK225254.1 | - | 1 | 799 | ||
AK291119.1 | - | 7 | 799 | ||
BC006095.1 | - | 1 | 789 | 2821364 | |
BC008850.2 | - | 1 | 826 | 4138113 | |
BC009599.1 | - | 1 | 801 | 4134557 | |
BC071671.1 | - | 1 | 782 | 4392472 | |
BC071894.1 | - | 1 | 786 | 2906310 | |
BC071895.1 | - | 1 | 784 | 6568654 | |
BC087837.1 | - | 1 | 785 | 6170371 | |
L16558.1 | - | 1 | 779 | ||
X52967.1 | - | 7 | 800 | ||
X57958.1 | - | 1 | 816 | ||
X57959.1 | - | 1 | 789 |