Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009968.3 | NP_034098.1 | 9 | 1471 | |
GenBank mRNA | AK133820.1 | - | 10 | 1571 | |
AK134281.1 | - | 9 | 956 | ||
BC003800.1 | - | 9 | 963 | 3492859 | |
BC043076.1 | - | 9 | 932 | 6404996 | |
D78646.1 | - | 6 | 498 | ||
S70056.1 | - | 9 | 983 |