Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011814.2 | NP_035944.2 | 17 | 2912 | |
GenBank mRNA | AK154243.1 | - | 17 | 2911 | |
AK192714.1 | - | 1 | 252 | ||
AK192912.1 | - | 1 | 254 | ||
AK195253.1 | - | 1 | 252 | ||
AK195317.1 | - | 1 | 252 | ||
BC057007.1 | - | 17 | 2807 | 6816943 | |
BC062971.1 | - | 17 | 2912 | 6415619 |