Gene Transcripts

Gene Symbol
Slc25a48
Entrez Gene ID
328258
Gene Name
solute carrier family 25, member 48
Gene Aliases
AU042651, E230025K15
Chromosome Location
Chr. 13: 56437963 - 56472367 on Build GRCm38
UniGene ID
Mm.482177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177809.4 NP_808477.2 5 736
GenBank mRNA AK054181.1 - 5 737
BC107250.1 - 5 700 40052351
BC107251.1 - 5 700 40052352
DV649007.1 - 2 347

View Full Product Details