Gene Transcripts
- Gene Symbol
- Krtap5-4
- Entrez Gene ID
- 50775
- Gene Name
- keratin associated protein 5-4
- Gene Aliases
- Chromosome Location
- Chr. 7: 142303502 - 142304503 on Build GRCm38
- UniGene ID
- Mm.389990
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015809.2 |
NP_056624.1
|
1 |
317 |
|
View Full Product Details