Gene Transcripts

Gene Symbol
UBAC1
Entrez Gene ID
10422
Gene Name
UBA domain containing 1
Gene Aliases
GBDR1, UBADC1
Chromosome Location
Chr. 9: 135932969 - 135961380 on Build GRCh38
UniGene ID
Hs.9194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016172.2 NP_057256.2 2 382
GenBank mRNA AF176796.1 - 2 355
BC004967.1 - 2 382 3535682
BC011822.1 - 2 208 3688526

View Full Product Details